XMD8-92
Lieferant: Apollo Scientific
Synonyme:
XMD8 92, 2-[2-ethoxy-4-(4-hydroxypiperidin-1-yl)anilino]-5,11-dimethylpyrimido[4,5-b][1,4]benzodiazepin-6-one, XMD8-92, XMD 8 92, XMD 892, XMD892, 2-((2-Ethoxy-4-(4-hydroxypiperidin-1-yl)phenyl)amino)-5,11-dimethyl-5,11-dihydro-6H-benzo[e]pyrimido[5,4-b][1,4]diazepin-6-one
BISN0197-1MG
APOSBISN0197-1MGEA
29
EUR
APOSBISN0197-1MG
XMD8-92
XMD8-92
MAPK/ERK Signalling Pathway
Formel:
C₂₆H₃₀N₆O₃ Molecular Weight: 474,55 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD18782742 CAS-Nummer: 1234480-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...