TAK-715
Lieferant: Apollo Scientific
Synonyme:
N-(4-(2-Ethyl-4-(m-tolyl)thiazol-5-yl)pyridin-2-yl)benzamide
BISN0206-1MG
APOSBISN0206-1MGEA
33.5
EUR
APOSBISN0206-1MG
TAK-715
TAK-715
MAPK/ERK Signalling Pathway
Formel:
C₂₄H₂₁N₃OS Molecular Weight: 399,5 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD17012805 CAS-Nummer: 303162-79-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...