Binimetinib
Lieferant: Apollo Scientific
Synonyme:
MEK-162, 5-((4-Bromo-2-fluorophenyl)amino)-4-fluoro-N-(2-hydroxyethoxy)-1-methyl-1H-benzo[d]imidazole-6-carboxamide
BISN0213-5MG
BISN0213-10MG
BISN0213-50MG
APOSBISN0213-5MGEA
79.9
EUR
APOSBISN0213-5MG
APOSBISN0213-10MG
APOSBISN0213-50MG
Binimetinib
Binimetinib
MAPK/ERK Signalling Pathway
Formel:
C₁₇H₁₅BrF₂N₄O₃ Molecular Weight: 441,23 g/mol Schmelzpunkt: 215…222 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD22124525 CAS-Nummer: 606143-89-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...