Pamapimod

Lieferant: Apollo Scientific

Synonyme: Ro-4402257, Ro 4402257, R1503, R-1503, Pamapimod, Ro4402257, 6-(2,4-Difluorophenoxy)-2-((1,5-dihydroxypentan-3-yl)amino)-8-methylpyrido[2,3-d]pyrimidin-7(8H)-one, R 1503

BISN0219-1MG BISN0219-5MG
APOSBISN0219-1MGEA 190 EUR
APOSBISN0219-1MG APOSBISN0219-5MG
Pamapimod
Pamapimod

MAPK/ERK Signalling Pathway

Formel: C₁₉H₂₀F₂N₄O₄
Molecular Weight: 406,38 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14105621
CAS-Nummer: 449811-01-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR