AZ-304
Lieferant: Apollo Scientific
Synonyme:
AZ304, 3-(2-Cyanopropan-2-yl)-N-(3-((7-methoxyquinazolin-4-yl)amino)-4-methylphenyl)benzamide
BISN0220-50MG
APOSBISN0220-50MGEA
89.9
EUR
APOSBISN0220-50MG
AZ-304
AZ-304
MAPK/ERK Signalling Pathway
Formel:
C₂₇H₂₅N₅O₂ Molecular Weight: 451,53 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31812677 CAS-Nummer: 942507-42-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...