SB239063
Lieferant: Apollo Scientific
Synonyme:
4-[4-(4-fluorophenyl)-5-(2-methoxypyrimidin-4-yl)imidazol-1-yl]cyclohexan-1-ol, trans-4-[4-(4-fluorophenyl)-5-(2-methoxy-4-pyrimidinyl)-1H-imidazol-1-yl]-cyclohexanol, Rel-(1r,4r)-4-(4-(4-fluorophenyl)-5-(2-methoxypyrimidin-4-yl)-1H-imidazol-1-yl)cyclohexan-1-ol, 4-(4-(4-fluorophenyl)-5-(2-methoxypyrimidin-4-yl)-1H-imidazol-1-yl)cyclohexanol
BISN0226-5MG
APOSBISN0226-5MGEA
81.2
EUR
APOSBISN0226-5MG
SB239063
SB239063
MAPK/ERK Signalling Pathway
Formel:
C₂₀H₂₁FN₄O₂ Molecular Weight: 368,4 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03792786 CAS-Nummer: 193551-21-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...