L-779450
Lieferant: Apollo Scientific
Synonyme:
2-Chloro-5-(2-phenyl-5-(pyridin-4-yl)-1H-imidazol-4-yl)phenol, Raf Kinase Inhibitor IV, L 779450, 2-Chloro-5-(2-phenyl-4-(pyridin-4-yl)-1H-imidazol-5-yl)phenol, 2-(Phenyl)-4-(3-hydroxy-4-chlorophenyl)-5-(4-pyridyl)-1H-imidazole, L779450
BISN0240-50MG
APOSBISN0240-50MGEA
119
EUR
APOSBISN0240-50MG
L-779450
L-779450
MAPK/ERK Signalling Pathway
Formel:
C₂₀H₁₄ClN₃O Molecular Weight: 347,80 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12828750 CAS-Nummer: 303727-31-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...