AZ 628
Lieferant: Apollo Scientific
Synonyme:
AZ 628, 3-(2-cyanopropan-2-yl)-N-[4-methyl-3-[(3-methyl-4-oxoquinazolin-6-yl)amino]phenyl]benzamide, AZ-628, AZ628, 3-(2-Cyanopropan-2-yl)-N-(4-methyl-3-((3-methyl-4-oxo-3,4-dihydroquinazolin-6-yl)amino)phenyl)benzamide
BISN0260-25MG
APOSBISN0260-25MGEA
148
EUR
APOSBISN0260-25MG
AZ 628
AZ 628
MAPK/ERK Signalling Pathway
Formel:
C₂₇H₂₅N₅O₂ Molecular Weight: 451,53 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD17392577 CAS-Nummer: 878739-06-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...