LJH685
Lieferant: Apollo Scientific
Synonyme:
2,6-difluoro-4-[4-[4-(4-methyl-1-piperazinyl)phenyl]-3-pyridinyl]-phenol, 2,6-Difluoro-4-(4-(4-(4-methylpiperazin-1-yl)phenyl)pyridin-3-yl)phenol
BISN0262-50MG
APOSBISN0262-50MGEA
188
EUR
APOSBISN0262-50MG
LJH685
LJH685
MAPK/ERK Signalling Pathway
Formel:
C₂₂H₂₁F₂N₃O Molecular Weight: 381,40 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29924738 CAS-Nummer: 1627710-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...