PD318088

Lieferant: Apollo Scientific

Synonyme: PD-318088, 5-Bromo-N-(2,3-dihydroxypropoxy)-3,4-difluoro-2-((2-fluoro-4-iodophenyl)amino)benzamide, pd 318088, PD318088, pd318088, PD 318088, 5-bromo-N-(2,3-dihydroxypropoxy)-3,4-difluoro-2-(2-fluoro-4-iodoanilino)benzamide

BISN0284-25MG
APOSBISN0284-25MGEA 128 EUR
APOSBISN0284-25MG
PD318088
PD318088

MAPK/ERK Signalling Pathway

Formel: C₁₆H₁₃BrF₃IN₂O₄
Molecular Weight: 561,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18384973
CAS-Nummer: 391210-00-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR