TA-02
Lieferant: Apollo Scientific
Synonyme:
4-(2-(2-fluorophenyl)-4-(4-fluorophenyl)-1H-imidazol-5-yl)pyridine
BISN0290-1MG
BISN0290-5MG
BISN0290-10MG
APOSBISN0290-1MGEA
31.9
EUR
APOSBISN0290-1MG
APOSBISN0290-5MG
APOSBISN0290-10MG
TA-02
TA-02
MAPK/ERK Signalling Pathway
Formel:
C₂₀H₁₃F₂N₃ Molecular Weight: 333,33 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29924749 CAS-Nummer: 1784751-19-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...