CH4987655

Lieferant: Apollo Scientific

Synonyme: RO4987655, 3,4-Difluoro-2-((2-fluoro-4-iodophenyl)amino)-N-(2-hydroxyethoxy)-5-((3-oxo-1,2-oxazinan-2-yl)methyl)benzamide

BISN0293-5MG BISN0293-10MG
APOSBISN0293-5MGEA 415 EUR
APOSBISN0293-5MG APOSBISN0293-10MG
CH4987655
CH4987655

MAPK/ERK Signalling Pathway

Formel: C₂₀H₁₉F₃IN₃O₅
Molecular Weight: 565,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19443685
CAS-Nummer: 874101-00-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR