FR180204

Lieferant: Apollo Scientific

Synonyme: FR180204, FR 180204, FR-180204, 5-(2-phenylpyrazolo[1,5-a]pyridin-3-yl)-2H-pyrazolo[3,4-c]pyridazin-3-amine, 5-(2-Phenylpyrazolo[1,5-a]pyridin-3-yl)-1H-pyrazolo[3,4-c]pyridazin-3-amine

BISN0306-1MG BISN0306-5MG BISN0306-10MG
APOSBISN0306-1MGEA 37.7 EUR
APOSBISN0306-1MG APOSBISN0306-5MG APOSBISN0306-10MG
FR180204
FR180204

MAPK/ERK Signalling Pathway

Formel: C₁₈H₁₃N₇
Molecular Weight: 327,34 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585805
CAS-Nummer: 865362-74-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR