Disulfiram

Lieferant: Apollo Scientific

Synonyme: Tetraethylthiuramdisulfid, Bis(diethyl-thiocarbamoyl)disulfid

BISN0322-1G BISN0322-5G
APOSBISN0322-1GEA 72.5 EUR
APOSBISN0322-1G APOSBISN0322-5G
Disulfiram
Disulfiram
Formel: C₁₀H₂₀N₂S₄
Molecular Weight: 296,55 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00009048
CAS-Nummer: 97-77-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR