Shikonin

Lieferant: Apollo Scientific

Synonyme: (R)-5,8-Dihydroxy-2-(1-hydroxy-4-methylpent-3-en-1-yl)naphthalene-1,4-dione, Tokyo Violet, 5,8-Dihydroxy-2-[(1R)-1-hydroxy-4-methyl-3-penten-1-yl]-1,4-naphthalenedione, C.I. 75535

BISN0323-10MG BISN0323-50MG BISN0323-100MG
APOSBISN0323-10MGEA 72.5 EUR
APOSBISN0323-10MG APOSBISN0323-50MG APOSBISN0323-100MG
Shikonin
Shikonin
Formel: C₁₆H₁₆O₅
Molecular Weight: 288,3 g/mol
Schmelzpunkt: 147 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00075680
CAS-Nummer: 517-89-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR