TDZD-8
Lieferant: Apollo Scientific
Synonyme:
4-Benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione
BISN0325-1MG
APOSBISN0325-1MGEA
24.8
EUR
APOSBISN0325-1MG
TDZD-8
TDZD-8
Formel:
C₁₀H₁₀N₂SO₂ Molecular Weight: 222,3 g/mol Schmelzpunkt: 63…64,4 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04973552 CAS-Nummer: 327036-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...