Tri-O-acetyl-D-galactal

Lieferant: Apollo Scientific
Achtung

Synonyme: (2R,3R,4R)-2-(Acetoxymethyl)-3,4-dihydro-2H-pyran-3,4-diyl diacetate, 1,5-Anhydro-2-deoxy-D-lyxo-hex-1-enitol 3,4,6-Tri-O-acetyl Ether, 3,4,6-Tri-O-acetyl-D-galactal

BISY010-10G BISY010-25G BISY010-100G
APOSBISY010-10GEA 108 EUR
APOSBISY010-10G APOSBISY010-25G APOSBISY010-100G
Tri-O-acetyl-D-galactal
Tri-O-acetyl-D-galactal
Formel: C₁₂H₁₆O₇
Molecular Weight: 272,25 g/mol
Schmelzpunkt: 34…38 °C
Dichte: 1,23 g/cm³ (25 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00064092
CAS-Nummer: 4098-06-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR