Trimethoprim 99,8%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(3,4,5-Trimethoxybenzyl)pyrimidine-2,4-diamine, 2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine

BIT0154-25G BIT0154-100G BIT0154-500G
APOSBIT0154-25GEA 21.9 EUR
APOSBIT0154-25G APOSBIT0154-100G APOSBIT0154-500G
Trimethoprim 99,8%
Trimethoprim

This antibiotic is a dihydrofolate reductase inhibitor. It inhibits the conversion of bacterial dihydrofolic acid to tetrahydrofolic acid which is necessary for the synthesis of amino acids, etc. and ultimately DNA synthesis.

Formel: C₁₄H₁₈N₄O₃
Molecular Weight: 290,32 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00036761
CAS-Nummer: 738-70-5
EINECS: 212-006-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR