Tiamulin fumarate

Lieferant: Apollo Scientific
Achtung

BIT0157-1G BIT0157-250MG BIT0157-5G
APOSBIT0157-1GEA 79.9 EUR
APOSBIT0157-1G APOSBIT0157-250MG APOSBIT0157-5G
Tiamulin fumarate
Tiamulin fumarate

  • Antibodies

Formel: C₃₂H₅₁NO₈S
Lagertemperatur: Kühlschrank
MDL: MFCD00145407
CAS-Nummer: 55297-96-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR