L-Threonin ≥99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid
BIT0719-100G
BIT0719-1KG
BIT0719-500G
APOSBIT0719-100GEA
21.9
EUR
APOSBIT0719-100G
APOSBIT0719-1KG
APOSBIT0719-500G
L-Threonin ≥99%
L-Threonin
USP, pyrogen free
Formel:
C₄H₉NO₃ Molecular Weight: 119.12 g/mol Schmelzpunkt: 255…256 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064270 CAS-Nummer: 72-19-5 EINECS: 200-774-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...