DL-Threonin

Lieferant: Apollo Scientific
Achtung

BIT0724-100G BIT0724-250G
APOSBIT0724-100GEA 125 EUR
APOSBIT0724-100G APOSBIT0724-250G
DL-Threonin
DL-Threonin
Formel: C₄H₉NO₃
Molecular Weight: 119.12 g/mol
Schmelzpunkt: 244 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00063722
CAS-Nummer: 80-68-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR