Thiolutin (aus Streptomyces luteosporeus)
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
6-(Acetamido)-4-methyl-1,2-dithiolo[4,3-b]-pyrrol-5(4H)-one
BIT1001-10MG
BIT1001-5MG
APOSBIT1001-10MGEA
480
EUR
APOSBIT1001-10MG
APOSBIT1001-5MG
Thiolutin (aus Streptomyces luteosporeus)
Thiolutin
Potent inhibitor of bacterial and yeast RNA polymerases.
Formel:
C₈H₈N₂O₂S₂ Schmelzpunkt: 281…285 °C Lagertemperatur: Gefrierschrank |
CAS-Nummer:
87-11-6 UN: 2811 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...