2,4,6-Tris(2-pyridyl)-1,3,5-triazin

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4,6-Tris(2-pyridyl)-s-triazin

BIT3089-10G BIT3089-25G BIT3089-5G
APOSBIT3089-10GEA 43.5 EUR
APOSBIT3089-10G APOSBIT3089-25G APOSBIT3089-5G
2,4,6-Tris(2-pyridyl)-1,3,5-triazin
2,4,6-Tris(2-pyridyl)-1,3,5-triazin
Formel: C₁₈H₁₂N₆
Molecular Weight: 312,33 g/mol
Schmelzpunkt: 247…249 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00006045
CAS-Nummer: 3682-35-7
EINECS: 222-965-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR