Zink 98%, Staub max. 10 μm

Lieferant: Apollo Scientific
Achtung

IN1013-100G IN1013-500G
APOSIN1013-100GEA 21.9 EUR
APOSIN1013-100G APOSIN1013-500G
Zink 98%, Staub max. 10 μm
Zinc

Delivery of this item to mainland UK is £40, additional charge will be added at order confirmation stage by our sales team.

Formel: Zn
Molecular Weight: 65.39 g/mol
Siedepunkt: 907 °C
Schmelzpunkt: 420 °C
Dichte: 7.133 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011291
CAS-Nummer: 7440-66-6
EINECS: 231-175-3
UN: 3077
ADR: 9+EHS III
Merck Index: 12,10255

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR