Dysprosium(III)bromid Hydrat 99,9%

Lieferant: Apollo Scientific

IN1633-100G IN1633-500G
APOSIN1633-100GEA 422 EUR
APOSIN1633-100G APOSIN1633-500G
Dysprosium(III)bromid Hydrat 99,9%
Dysprosium(III)bromid Hydrat
Formel: Br₃Dy·H₂O
Schmelzpunkt: 881 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 29843-91-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR