Erbiumtribromid Nonahydrat 99.99%
Lieferant: Apollo Scientific
Synonyme:
Erbium (III) bromide nonahydrate
IN1687-100G
IN1687-25G
APOSIN1687-100GEA
371
EUR
APOSIN1687-100G
APOSIN1687-25G
Erbiumtribromid Nonahydrat 99.99%
Erbiumtribromid Nonahydrat
Formel:
Br₃Er·9H₂O Molecular Weight: 569,11 g/mol Schmelzpunkt: 950 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00209649 CAS-Nummer: 29843-93-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...