Europiumtrinitrat Hydrat 99,9%
Lieferant: Apollo Scientific
|
Gefahr
|
IN1777-100G
IN1777-10G
APOSIN1777-100GEA
0
EUR
APOSIN1777-100G
APOSIN1777-10G
Europiumtrinitrat Hydrat 99,9%
Europiumtrinitrat Hydrat
Formel:
EuN₃O₉·xH₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149697 CAS-Nummer: 100587-95-9 UN: 1477 ADR: 5.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...