Europiumtrinitrat Hydrat 99.99%

Lieferant: Apollo Scientific
Achtung

Synonyme: Europium (III) nitrate hydrate

IN1780-25G IN1780-5G IN1780-100G
APOSIN1780-25GEA 574 EUR
APOSIN1780-25G APOSIN1780-5G APOSIN1780-100G
Europiumtrinitrat Hydrat 99.99%
Europiumtrinitrat Hydrat
Formel: EuN₃O₉·xH₂O
Molecular Weight: 337,98 g/mol (anhy)
Schmelzpunkt: 85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149697
CAS-Nummer: 100587-95-9
UN: 1477
ADR: 5.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR