Europiumtrinitrat Hydrat 99.999%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Europium (III) nitrate hydrate
IN1783-25G
IN1783-5G
IN1783-100G
APOSIN1783-25GEA
509
EUR
APOSIN1783-25G
APOSIN1783-5G
APOSIN1783-100G
Europiumtrinitrat Hydrat 99.999%
Europiumtrinitrat Hydrat
Formel:
EuN₃O₉·xH₂O Molecular Weight: 337,98 g/mol (anhy) Schmelzpunkt: 85 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149697 CAS-Nummer: 100587-95-9 UN: 1477 ADR: 5.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...