Lanthanum(III) nitrate hydrate 99.99%

Lieferant: Apollo Scientific

IN2152-100G IN2152-500G
APOSIN2152-100GEA 72.5 EUR
APOSIN2152-100G APOSIN2152-500G
Lanthanum(III) nitrate hydrate 99.99%
Lanthanum(III) nitrate hydrate
Formel: H₂LaN₃O₁₀
Molecular Weight: 342,932 g/mol
MDL: MFCD00011067
CAS-Nummer: 100587-94-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR