Neodymium(III) acetate hydrate 99,9%

Lieferant: Apollo Scientific
Achtung

IN2623-50G IN2623-250G
APOSIN2623-50GEA 109 EUR
APOSIN2623-50G APOSIN2623-250G
Neodymium(III) acetate hydrate 99,9%
Neodymium(III) acetate hydrate
Formel: C₆H₁₁NdO₇
Lagertemperatur: Raumtemperatur
MDL: MFCD00150120
CAS-Nummer: 334869-71-5
EINECS: 228-244-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR