Tris(dibenzylideneacetone)dipalladium(0)

Lieferant: Apollo Scientific
Achtung

Synonyme: Pd2dba3, Pd₂(dba)₃

IN2823-1G IN2823-25G IN2823-5G
APOSIN2823-1GEA 31.9 EUR
APOSIN2823-1G APOSIN2823-25G APOSIN2823-5G
Tris(dibenzylideneacetone)dipalladium(0)
Tris(dibenzylideneacetone)dipalladium(0)
Formel: C₅₁H₄₂O₃Pd₂
Molecular Weight: 915,72 g/mol
Schmelzpunkt: 152…155 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00013310
CAS-Nummer: 51364-51-3
EINECS: 479-280-5
UN: 3077
ADR: 9+EHS,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR