Potassium trimethylsilanolate 90%

Lieferant: Apollo Scientific
Gefahr

Synonyme: trimethylsiloxypotassium, potassium and trimethyl(oxido)silane, Silanol, 1,1,1-trimethyl-, potassium salt (1:1)

IN2946-100G IN2946-25G
APOSIN2946-100GEA 186 EUR
APOSIN2946-100G APOSIN2946-25G
Potassium trimethylsilanolate 90%
Potassium trimethylsilanolate
Formel: C₃H₉KOSi
Molecular Weight: 128,29 g/mol
Schmelzpunkt: 135…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013421
CAS-Nummer: 10519-96-7
UN: 3263
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR