Scandium(III)chlorid Hexahydrat 99.99%

Lieferant: Apollo Scientific
Achtung

IN3130-1G IN3130-5G
APOSIN3130-1GEA 232 EUR
APOSIN3130-1G APOSIN3130-5G
Scandium(III)chlorid Hexahydrat 99.99%
Scandium(III)chlorid Hexahydrat
Formel: Cl₃Sc·6H₂O
Molecular Weight: 259,41 g/mol
Schmelzpunkt: 63…66 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149852
CAS-Nummer: 20662-14-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR