Terbium(III)fluorid 99,9%
Lieferant: Apollo Scientific
Synonyme:
Terbiumtrifluorid
IN3478-25G
IN3478-5G
APOSIN3478-25GEA
343
EUR
APOSIN3478-25G
APOSIN3478-5G
Terbium(III)fluorid 99,9%
Terbium(III)fluorid
Formel:
TbF₃ Schmelzpunkt: 1172 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00049572 CAS-Nummer: 13708-63-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...