Terbium(III,IV)oxid 99,9%

Lieferant: Apollo Scientific
Achtung

Synonyme: Tetraterbium heptaoxide, Terbium oxide

IN3487-25G IN3487-5G IN3487-100G
APOSIN3487-25GEA 261 EUR
APOSIN3487-25G APOSIN3487-5G APOSIN3487-100G
Terbium(III,IV)oxid 99,9%
Terbium(III,IV)oxid
Formel: O₇Tb₄
Molecular Weight: 747.7 g/mol
Schmelzpunkt: 2340 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00083151
CAS-Nummer: 12037-01-3
EINECS: 234-856-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR