Terbium(III)sulfat Octahydrat 99,9%
Lieferant: Apollo Scientific
Synonyme:
Diterbiumtrisulfat Octahydrat
IN3493-10G
APOSIN3493-10GEA
306
EUR
APOSIN3493-10G
Terbium(III)sulfat Octahydrat 99,9%
Terbium(III)sulfat Octahydrat
Formel:
O₁₄S₃Tb₂·8H₂O Molecular Weight: 782,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149872 CAS-Nummer: 13842-67-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...