Thulium(III)chlorid, wasserfrei 99,9%

Lieferant: Apollo Scientific
Achtung

IN3601-1G IN3601-5G
APOSIN3601-1GEA 174 EUR
APOSIN3601-1G APOSIN3601-5G
Thulium(III)chlorid, wasserfrei 99,9%
Thulium(III)chlorid
Formel: Cl₃Tm
Molecular Weight: 275,29 g/mol
Schmelzpunkt: 821 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011283
CAS-Nummer: 13537-18-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR