2'-Aminoacetanilid

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(2-Aminophenyl)acetamid

OR0008-100G OR0008-25G OR0008-5G
APOSOR0008-100GEA 161 EUR
APOSOR0008-100G APOSOR0008-25G APOSOR0008-5G
2'-Aminoacetanilid
2'-Aminoacetanilid
Formel: C₈H₁₀N₂O
Molecular Weight: 150,18 g/mol
Schmelzpunkt: 133…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00210388
CAS-Nummer: 34801-09-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR