N-(4-Bromphenyl)maleimid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(4-Bromphenyl)maleinsäureimid

APOSOR0012-25GEA 115 EUR
APOSOR0012-25G APOSOR0012-1G APOSOR0012-5G
N-(4-Bromphenyl)maleimid 98%
N-(4-Bromphenyl)maleimid
Formel: C₁₀H₆BrNO₂
Molecular Weight: 252,07 g/mol
Schmelzpunkt: 135…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00030664
CAS-Nummer: 13380-67-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR