Bis(4-chlorphenyl)disulfid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Bis(4-chlorophenyl) disulphide
OR0016-10G
OR0016-50G
APOSOR0016-10GEA
26.1
EUR
APOSOR0016-10G
APOSOR0016-50G
Bis(4-chlorphenyl)disulfid 98%
Bis(4-chlorphenyl)disulfid
Formel:
C₁₂H₈Cl₂S₂ Molecular Weight: 287.23 g/mol Schmelzpunkt: 71…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013642 CAS-Nummer: 1142-19-4 EINECS: 214-531-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...