5-Brom-2-chlorphenol 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR0018-25G
OR0018-5G
OR0018-100G
APOSOR0018-25GEA
55.1
EUR
APOSOR0018-25G
APOSOR0018-5G
APOSOR0018-100G
5-Brom-2-chlorphenol 98%
5-Brom-2-chlorphenol
Formel:
C₆H₄BrClO Molecular Weight: 207,45 g/mol Siedepunkt: 234 °C Schmelzpunkt: 55…58 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00672940 CAS-Nummer: 183802-98-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...