4'-Acetamido-2'-methylacetophenon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(4-Acetyl-3-methylphenyl)acetamide
OR0064-1G
OR0064-5G
APOSOR0064-1GEA
29
EUR
APOSOR0064-1G
APOSOR0064-5G
4'-Acetamido-2'-methylacetophenon
4'-Acetamido-2'-methylacetophenon
Formel:
C₁₁H₁₃NO₂ Molecular Weight: 191.23 g/mol Schmelzpunkt: 137…140 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02258876 CAS-Nummer: 34956-31-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...