Ethyl-2-amino-5-methyl-4-phenyl-3-thiophencarboxylat 95%

Lieferant: Apollo Scientific
Achtung

OR0092-1G OR0092-250MG OR0092-5G
APOSOR0092-1GEA 0 EUR
APOSOR0092-1G APOSOR0092-250MG APOSOR0092-5G
Ethyl-2-amino-5-methyl-4-phenyl-3-thiophencarboxylat 95%
Ethyl-2-amino-5-methyl-4-phenyl-3-thiophencarboxylat
Formel: C₁₄H₁₅NO₂S
Molecular Weight: 261.34 g/mol
Schmelzpunkt: 72…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01763475
CAS-Nummer: 4815-37-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR