2-Benzo[b]thiophenboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: Benzo[b]thiophen-2-ylboronic acid, 2-Benzothienylboronic acid

OR0093-1G OR0093-25G OR0093-5G
APOSOR0093-1GEA 21.9 EUR
APOSOR0093-1G APOSOR0093-25G APOSOR0093-5G
2-Benzo[b]thiophenboronsäure
2-Benzo[b]thiophenboronsäure
Formel: C₈H₇BO₂S
Molecular Weight: 178.02 g/mol
Schmelzpunkt: 252…255 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD01075674
CAS-Nummer: 98437-23-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR