Ethyl(3,4-dichlorbenzoyl)formiat 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl (3,4-dichlorophenyl)(oxo)acetate

OR0096-1G OR0096-5G
APOSOR0096-1GEA 55.1 EUR
APOSOR0096-1G APOSOR0096-5G
Ethyl(3,4-dichlorbenzoyl)formiat 95%
Ethyl(3,4-dichlorbenzoyl)formiat
Formel: C₁₀H₈Cl₂O₃
Molecular Weight: 247,08 g/mol
Siedepunkt: 121…123 °C/249mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD01934864
CAS-Nummer: 34966-52-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR