5-Brompyrimidin 98%

Lieferant: Apollo Scientific
Achtung

OR0101-100G OR0101-250G
APOSOR0101-100GEA 132 EUR
APOSOR0101-100G APOSOR0101-250G
5-Brompyrimidin 98%
5-Bromopyrimidine
Formel: C₄H₃BrN₂
Molecular Weight: 158,99 g/mol
Schmelzpunkt: 69…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006117
CAS-Nummer: 4595-59-9
EINECS: 224-992-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR