3-Brombenzolsulfonamid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Brombenzolsulfonsäureamid

OR0112-1G OR0112-5G
APOSOR0112-1GEA 21.9 EUR
APOSOR0112-1G APOSOR0112-5G
3-Brombenzolsulfonamid
3-Brombenzolsulfonamid
Formel: C₆H₆BrNO₂S
Molecular Weight: 236,09 g/mol
Schmelzpunkt: 154…158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00084903
CAS-Nummer: 89599-01-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR