2-Anilinonicotinic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Phenylamino)nicotinic acid

OR01497-500MG OR01497-1G
APOSOR01497-500MGEA 52.2 EUR
APOSOR01497-500MG APOSOR01497-1G
2-Anilinonicotinic acid
2-Anilinonicotinic acid
Formel: C₁₂H₁₀N₂O₂
Molecular Weight: 214.23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00128791
CAS-Nummer: 16344-24-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR