Ethyl-2-(hydroxyimino)-3-oxobutanoate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-(hydroxyimino)-3-oxobutanoate

OR015018-1G OR015018-25G OR015018-5G OR015018-250MG OR015018-500MG OR015018-10G
APOSOR015018-1GEA 81.2 EUR
APOSOR015018-1G APOSOR015018-25G APOSOR015018-5G APOSOR015018-250MG APOSOR015018-500MG APOSOR015018-10G
Ethyl-2-(hydroxyimino)-3-oxobutanoate
Ethyl-2-(hydroxyimino)-3-oxobutanoate
Formel: C₆H₉NO₄
Molecular Weight: 159,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00039517
CAS-Nummer: 5408-04-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR